Learn More
ITGA2 Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547193
264.01 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, approximately10% cross-reactivity with recombinant human Integrin alpha M is observed and less than 1% cross-reactivity with recombinant mouse (rm) Integrin alpha 5 and rmIntegrin alpha E is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ITGA2 Polyclonal antibody specifically detects ITGA2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Specifications
ITGA2 | |
Polyclonal | |
Unconjugated | |
Itga2 | |
alpha 2 subunit of VLA-2 receptor; BDPLT9; BR; CD49 antigen-like family member B; CD49B; Collagen receptor; DX5; GPIa; HPA-5; human platelet alloantigen system 5; integrin alpha 2; integrin alpha-2; integrin subunit alpha 2; integrin, alpha 2; integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor); Itga2; platelet antigen Br; platelet glycoprotein GPIa; platelet membrane glycoprotein Ia; very late activation protein 2 receptor, alpha-2 subunit; VLA-2; VLA2 receptor; VLA-2 receptor, alpha 2 subunit; VLA-2 subunit alpha; VLAA2 | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
16398, 3673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P17301, Q62469 | |
Itga2 | |
Mouse myeloma cell line NS0-derived recombinant mouse Integrin alpha 2/CD49b Tyr27-Thr1129 | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |