Learn More
Integrin alpha 2/CD49b Rat anti-Human, Clone: 430907, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB12331
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438475
  • £249.49 / 100µg
Estimated Shipment date
from Supplier 27-09-2019
Description
Description
Integrin alpha 2/CD49b Monoclonal antibody MAB12331 Specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Western Blotting
Specifications
Specifications
Integrin alpha 2/CD49b
430907
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ITGA2
Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 2/CD49b Tyr30-Thr1132 Accession # P17301
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3673
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P17301
alpha-2 subunit, CD49b, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), ITGA2, VLA 2, VLA-2 alpha
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Integrin alpha 2/CD49b in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant mouse (rm) Integrin alpha 2 is observed and no cross-reactivity with recombinant human (rh) Integrin alpha 1, alpha 3, alpha 5, alpha 6, alpha 8, alpha 9, alpha 10, alpha 11, alpha L, alpha M, or alpha V is observed.