missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iNOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517106
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
iNOS Polyclonal antibody specifically detects iNOS in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
iNOS | |
Polyclonal | |
Unconjugated | |
Nos2 | |
Hepatocyte NOS; hepatocytes; HEP-NOS; inducible nitric oxide synthase; inducible NO synthase; Inducible NOS; iNos; i-NOS; Inosl; MAC-NOS; macrophage NOS; nitric oxide synthase 2; nitric oxide synthase 2, inducible; nitric oxide synthase 2, inducible, macrophage; nitric oxide synthase 2A (inducible, hepatocytes); nitric oxide synthase, inducible; nitric oxide synthase, macrophage; nitric oxide synthase-inducible; NOS; NOS type II; NOS, type II; Nos2; Nos-2; Nos2a; NOS-II; OTTMUSP00000000202; peptidyl-cysteine S-nitrosylase NOS2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18126, 24599, 4843 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
19 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P29477, P35228, Q06518 | |
Nos2 | |
Synthetic peptide sourrounding Ala1130 of mouse iNOS | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |