missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A NS1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523365
Code : LD
Additional Details : Weight : 0.01000kg
Description
Influenza A NS1 Polyclonal antibody specifically detects Influenza A NS1 in Virus samples. It is validated for Western Blot.Specifications
Influenza A NS1 | |
Polyclonal | |
Unconjugated | |
Influenza NS1 | |
Within the range of amino acids 24-84 of human Influenza A H1N1 nonstructural protein | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Virus | |
Antibody | |
IgG |