missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Influenza A H1N1 NA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523412
Code : LD
Additional Details : Weight : 0.01000kg
Description
Influenza A H1N1 NA Polyclonal antibody specifically detects Influenza A H1N1 NA in Virus samples. It is validated for Western Blot.Specifications
Influenza A H1N1 NA | |
Polyclonal | |
Unconjugated | |
H1N1 NA; Influenza H1N1 | |
Within the range of amino acids 35-58 of Influenza A H1N1 NA (swine flu) | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Rabbit | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG |