missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524598
Code : LD
Additional Details : Weight : 0.01000kg
Description
IDO Polyclonal antibody specifically detects IDO in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
IDO | |
Polyclonal | |
Unconjugated | |
Ido1 | |
EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
15930, 3620 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
PBS with 0.09% sodium azide | |
P14902, P28776 | |
Ido1 | |
KLH conjugated synthetic peptide between 79-105 amino acids from the central region of human INDO | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |