missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDO Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA512305
Code : LE
Additional Details : Weight : 0.01000kg
Description
IDO Polyclonal antibody specifically detects IDO in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
IDO | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P14902 | |
Ido1 | |
KLH conjugated synthetic peptide between 374-403 amino acids from the C-terminal region of human INDO | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Ido1 | |
EC 1.13.11.52; IDO; IDO1; IDO-1; INDO; indolamine 2,3 dioxygenase; indole 2,3-dioxygenase; indoleamine; indoleamine 2,3-dioxygenase 1; indoleamine 23-dioxygenase; Indoleamine-2; indoleamine-pyrrole 2,3 dioxygenase; indoleamine-pyrrole 2,3-dioxygenase | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
3620 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |