Learn More
IL-6 R alpha Rat anti-Mouse, Clone: 255821, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1830
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458955
  • £337.93 / 500µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
IL-6 R alpha Monoclonal antibody MAB1830 Specifically detects IL-6 R alpha in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry
Specifications
Specifications
IL-6 R alpha
255821
Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready
P22272
CD126, gp80, IL6Q, IL6R, IL6R alpha, IL-6R-1, IL6RA, IL-6RA, IL6RQ, interleukin 6 receptor
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-6 R alpha in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-6 R alpha is observed.
Mouse
Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Il6ra
Mouse myeloma cell line NS0-derived recombinant mouse IL-6 R alpha Leu20-Glu357 Accession # P22272
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3570
Reconstitute at 0.5 mg/mL in sterile PBS.