Learn More
IL-6 Monoclonal Antibody (53325), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523749
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In sandwich ELISAs, shows less than 1% cross-reactivity with recombinant mouse IL-6, recombinant human IL-6, and recombinant porcine IL-6. Reconstitute at 0.5 mg/mL in sterile PBS.
IL-6 Monoclonal antibody specifically detects IL-6 in Rat samples. It is validated for ELISA.Specifications
IL-6 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P20607 | |
Il6 | |
E. coli-derived recombinant rat IL-6 Phe25-Thr211 | |
500 μg | |
Primary | |
Rat | |
Antibody | |
IgG2b |
ELISA | |
53325 | |
Unconjugated | |
Il6 | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Mouse | |
Protein A/G | |
RUO | |
24498 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |