Learn More
IL-5 Mouse anti-Rat, Clone: 119905, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB545
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460956
  • £337.93 / 500µg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
IL-5 Monoclonal antibody MAB545 Specifically detects IL-5 in Rat samples. It is validated for Western Blotting
Specifications
Specifications
IL-5
119905
Western Blot 1 ug/mL
Q08125
BCDF mu, B-cell differentiation factor I, BCGFII, EDF, Eo-CSF, Eosinophil differentiation factor, IL5, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRF, TRFB cell differentiation factor I
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat IL-5 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-5 and recombinant mouse IL-5 is observed.
Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL5
S. frugiperda insect ovarian cell line Sf 21-derived recombinant rat IL-5 Met20-Val132 Accession # Q08125
Protein A or G purified from ascites
RUO
Primary
3567
Reconstitute at 0.5 mg/mL in sterile PBS.