Learn More
IL4R Monoclonal Antibody (25463), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523814
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-9 R, recombinant mouse IL-4 R, rhIL-5 R alpha, rhIL-5 R beta, rhIL-13 R alpha 1, or rhIL-13 R alpha 2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL4R Monoclonal antibody specifically detects IL4R in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization, Western Blot.Specifications
IL4R | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P24394 | |
IL4R | |
Sf 21-derived recombinant human IL-4 R Gly24-His232 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization, Western Blot | |
25463 | |
Unconjugated | |
IL4R | |
582J2.1; CD124; il-4 receptor; IL-4 receptor alpha chain; IL-4 receptor subunit alpha; IL-4-binding protein; IL4-BP; Il4r; IL4R nirs variant 1; IL-4R subunit alpha; IL4RA; IL-4RA; IL-4R-alpha; interleukin 4 receptor; interleukin 4 receptor, alpha; interleukin-4 receptor alpha chain; interleukin-4 receptor subunit alpha; Soluble IL-4 receptor subunit alpha; Soluble interleukin-4 receptor subunit alpha | |
Mouse | |
Protein A/G | |
RUO | |
3566 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |