missing translation for 'onlineSavingsMsg'
Learn More

IL-4 R alpha Rat anti-Mouse, Clone: 129801, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB530

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460896

  • £373.57 / 500µg


Estimated Shipment
07-03-2019
Add to basket

Description

Description

IL-4 R alpha Monoclonal antibody MAB530 Specifically detects IL-4 R alpha in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-4 R alpha
129801
Western Blot 1 ug/mL
P16382
CD124, CD124 antigen, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-4 R alpha in direct ELISAs and Western blots. Shows less than 1% cross-reactivity with recombinant human IL-4 R alpha .
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Il4ra
Mouse myeloma cell line NS0-derived recombinant mouse IL-4 R alpha Ile26-Arg233 Accession # P16382
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3566
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only