missing translation for 'onlineSavingsMsg'
Learn More

IL-38/IL-1F10 Rat anti-Human, Clone: 316709, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2427

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449435

  • £311.64 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-38/IL-1F10 Monoclonal antibody MAB2427 Specifically detects IL-38/IL-1F10 in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-38/IL-1F10
316709
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL1F10
E. coli-derived recombinant human IL-38/IL-1F10 Met1-Trp152 Accession # Q8WWZ1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84639
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q8WWZ1
FIL1- theta, FKSG75, IL1F10, IL-1HY2, IL38, interleukin 1 family, member 10 (theta), UNQ6119/PRO20041
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-38/IL-1F10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse IL-1F10 is observed and no cross-reactivity with recombinant human (rh) IL-1 alpha/IL-1F1 rhIL-1 beta/IL-1F2, rhIL-1ra/IL-1F3, rhIL-18/IL-1F4, rhIL-36Ra/IL-1F5, rhIL-36 alpha/IL-1F6, rhIL-1F7/FIL1 zeta , rhIL-36 beta/IL-1F8, or rhIL-36 gamma/IL-1F9 is observed.
Human
Videos
Documents

For Research Use Only