Learn More
IL-36 alpha/IL-1F6 Mouse anti-Human, Clone: 162122, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1078
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438315
  • £337.93 / 500µg
Estimated Shipment
27-06-2019
Description
Description
IL-36 alpha/IL-1F6 Monoclonal antibody MAB1078 Specifically detects IL-36 alpha/IL-1F6 in Human samples. It is validated for Western Blotting
Specifications
Specifications
IL-36 alpha/IL-1F6
162122
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL36A
E. coli-derived recombinant human IL-36 alpha/IL-1F6 Met1-Phe158 Accession # Q9UHA7
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
27179
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9UHA7
FIL1, FIL1 epsilon, FIL1(EPSILON), FIL1E, IL-1 epsilon, IL1(EPSILON), IL1E, IL-1E, IL1F6, IL-1F6, IL-1F6 (FIL-1-epsilon), IL36 alpha, IL36A, interleukin 1 family, member 6 (epsilon), interleukin 1, epsilon, Interleukin 36, Alpha, Interleukin-1 epsilon, interleukin-1 family member 6, Interleukin-36 Alpha
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-36 alpha/IL-1F6 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-1 alpha , rhIL-1 beta , rhIL-1 receptor antagonist, rhIL-18, rhIL-36Ra, rhIL-36 beta or recombinant mouse IL-36 alpha is observed.