Learn More
IL-31 Rat anti-Mouse, Clone: 323407, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3028
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13429845
  • £313.11 / 100µg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
IL-31 Monoclonal antibody MAB3028 Specifically detects IL-31 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
IL-31
323407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL31
E. coli-derived recombinant mouse IL-31 Thr24-Cys163 Accession # Q6EAL8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
386653
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q6EAL8
IL31, IL-31interleukin-31, interleukin 31
Rat
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-31 in direct ELISAs and Western blots. Does not cross-react with recombinant human IL-31.