missing translation for 'onlineSavingsMsg'
Learn More

CD123 Monoclonal Antibody (32703), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA523818

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15513512

  • £374.00 / 100µg
Estimated Shipment: 08-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

CD123 Monoclonal antibody specifically detects CD123 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization, Western Blot.
Specifications

Specifications

CD123
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P26951
IL3RA
Sf 21-derived recombinant human IL-3R alpha/CD123 Lys20-Arg305, predicted
100 μg
Primary
Human
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Neutralization, Western Blot
32703
Unconjugated
IL3RA
CD123; CD123 antigen; CDw123; Cyrl; cytokine receptor-like protein CYRL; hIL-3Ra; Il-3 alpha subunit; IL-3 receptor alpha chain; IL-3 receptor alpha SP2 isoform; IL-3 receptor subunit alpha; IL3R; IL-3R subunit alpha; Il3ra; IL-3RA; IL-3R-alpha; IL3RAY; IL3RX; IL3RY; interleukin 3 receptor subunit alpha; interleukin 3 receptor, alpha; interleukin 3 receptor, alpha (low affinity); interleukin 3 receptor, alpha chain; interleukin-3 receptor class II alpha chain; Interleukin-3 receptor subunit alpha; Sut-1
Mouse
Protein A/G
RUO
3563
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers