Learn More
IL-3 R beta Rat anti-Mouse, Clone: 130710, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5491
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440976
  • £337.93 / 500µg
Estimated Shipment
09-07-2019
Description
Description
IL-3R beta Monoclonal antibody specifically detects IL-3R beta in Mouse samples. It is validated for ELISA.
Specifications
Specifications
IL-3R beta
130710
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P26954
AIC2A, BetaIl3, Bil3, Csfgmrb, IL3R, IL3R beta, IL-3Rb, IL3RB, Il3rb2
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-3 R beta in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse IL-3 R alpha or recombinant human IL-3 R alpha is observed. Cross-reactivity with AIC2B was not tested.
Mouse
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Csf2rb2
Mouse myeloma cell line NS0-derived recombinant mouse IL-3 R beta His23-Trp440 Accession # P26954
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1439
Reconstitute at 0.5 mg/mL in sterile PBS.