Learn More
IL-3 R alpha/CD123 Rat anti-Mouse, Clone: 151231, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB983
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13462066
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
IL-3 R alpha/CD123 Monoclonal antibody MAB983 Specifically detects IL-3 R alpha/CD123 in Mouse samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
IL-3 R alpha/CD123
151231
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
P26952
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL3R alpha, IL-3R subunit alpha, IL3RA, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-3 R alpha/CD123 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human IL-3 R alpha and recombinant mouse IL-3 R beta is observed.
Mouse
Western Blot, Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL3RA
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-3 R alpha/CD123 Ser17-Lys331 Accession # P26952
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3563
Reconstitute at 0.5 mg/mL in sterile PBS.