missing translation for 'onlineSavingsMsg'
Learn More

IL-3, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA175208

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11514330

  • £1112.86 / 500µg


Estimated Shipment
27-06-2018
Add to basket

Description

Description

PA1-75208 specifically detects IL-3.PA1-75208 has been successfully used in ELISA applications.The PA1-75208 immunogen is Recombinant IL-3.
Specifications

Specifications

IL-3
Polyclonal
PBS with 0.09% sodium azide
P08700
IL-3, MCGF, MGC79398, MGC79399, MULTI-CSF
Sheep
IgG
500μg
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
3562
ELISA
Unconjugated
IL3
Purified
IL3
Recombinant human IL-3
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos