missing translation for 'onlineSavingsMsg'
Learn More

IL-27 R alpha/WSX-1/TCCR Rat anti-Mouse, Clone: 263517, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB2109

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459185

  • £249.49 / 100µg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

IL-27 R alpha/WSX-1/TCCR Monoclonal antibody MAB2109 Specifically detects IL-27 R alpha/WSX-1/TCCR in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-27 R alpha/WSX-1/TCCR
263517
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL27RA
Mouse myeloma cell line NS0-derived recombinant mouse IL-27 R alpha/WSX-1/TCCR, Gly29-Lys510, Accession # O70394
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9466
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
O70394
class I cytokine receptor, CRL1IL-27R, Cytokine receptor-like 1, IL-27 R alpha, IL27R, IL27R alpha, IL27RA, IL-27RA, IL-27R-alpha, interleukin 27 receptor, alpha, interleukin-27 receptor subunit alpha, TCCR, TCCRIL-27 receptor subunit alpha, T-cell cytokine receptor type 1, Type I T-cell cytokine receptor, WSX-1, WSX1IL-27R subunit alpha, zcytor1
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-27 R alpha/WSX-1/TCCR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse gp130 or recombinant human IL-27 R alpha is observed.
Mouse
Videos

For Research Use Only