Learn More
IL-21 R Rat anti-Mouse, Clone: 155516, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5961
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13451156
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
IL-21R Monoclonal antibody specifically detects IL-21R in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
IL-21R
155516
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL21R
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse IL-21 R Cys20-Pro236 Accession # Q9JHX3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
50615
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9JHX3
CD360, CD360 antigen, IL-21 R, IL-21 receptor, IL21R, IL-21R, interleukin 21 receptor, interleukin-21 receptor, MGC10967, NILR, Novel interleukin receptor
Rat
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-21 R in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human IL-21 R or recombinant mouse IL-2 R beta is observed.