Learn More
IL-20 Rat anti-Mouse, Clone: 176007, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB12042
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438445
  • £337.93 / 500µg
Estimated Shipment
27-06-2019
Description
Description
IL-20 Monoclonal antibody MAB12042 Specifically detects IL-20 in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
IL-20
176007
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL20
E. coli-derived recombinant mouse IL-20 Leu25-Leu176 Accession # Q9JKV9.1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
50604
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q9JKV9.1
Cytokine Zcyto10, IL10D, IL20, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-20 in ELISAs.