missing translation for 'onlineSavingsMsg'
Learn More

IL-2 R beta Rat anti-Mouse, Clone: 130128, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB589

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441126

  • £390.72 / 500µg

Estimated Shipment date
from Supplier 06-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-2 R beta Monoclonal antibody MAB589 Specifically detects IL-2 R beta in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-2 R beta
130128
Western Blot 1 ug/mL
P16297
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-15 R beta, IL-2 receptor subunit beta, IL2R beta, IL-2R subunit beta, IL2RB, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-2 R beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IL-2 R beta , rhIL-2 R alpha , recombinant mouse (rm) IL-2 R gamma , rmIL-21 R, or rhIL-15 R is observed. In Western blots under reducing conditions, approximately 50% cross-reactivity with rhIL-2 R beta is observed.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL2RB
Mouse myeloma cell line NS0-derived recombinant mouse IL-2 R beta Ala27-Glu240 Accession # P16297
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3560
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only