Learn More
IL-2 Mouse anti-Cotton Rat, Clone: 144614, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB579
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441076
  • £337.93 / 500µg
Estimated Shipment
08-07-2019
Description
Description
IL-2 Monoclonal antibody MAB579 Specifically detects IL-2 in Cotton Rat samples. It is validated for Western Blotting
Specifications
Specifications
IL-2
144614
Western Blot 1 ug/mL
AF398549
aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects cotton rat IL-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 50-100% cross-reactivity with recombinant rat IL-2 is observed and no cross-reactivity with recombinant human IL-2, recombinant mouse (rm) IL-2, recombinant porcine IL-2, or rmIL-15 is observed.
Cotton Rat
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL2
E. coli-derived recombinant cotton rat IL-2 Ala21-Gln155 Accession # AF398549
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3558
Reconstitute at 0.5 mg/mL in sterile PBS.