Learn More
IL-2 Rat anti-Mouse, Clone: 142206, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB402
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440366
  • £337.93 / 500µg
Estimated Shipment
08-07-2019
Description
Description
IL-2 Monoclonal antibody MAB402 Specifically detects IL-2 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
IL-2
142206
Western Blot 1 ug/mL
P04351
aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-2 in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human (rh) IL-2, rrIL-2, rpIL-2, rcrIL-2, or rhIL-15.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL2
E. coli-derived recombinant mouse IL-2 Ala21-Gln169 Accession # P04351
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3558
Reconstitute at 0.5 mg/mL in sterile PBS.