missing translation for 'onlineSavingsMsg'
Learn More

IL-18 BPa Mouse anti-Human, Clone: 136031, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1192

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438435

  • £364.33 / 500µg

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-18 BPa Monoclonal antibody MAB1192 Specifically detects IL-18 BPa in Human samples. It is validated for ELISA Capture
Specifications

Specifications

IL-18 BPa/IL18BP
136031
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL18BP
Mouse myeloma cell line NS0-derived recombinant human IL-18 BPa Thr31-Gly194 (predicted) Accession # O95998
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10068
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
O95998
IL18 BPa, IL18BP, IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-18 BPa in ELISAs. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human (rh) IL-18, recombinant mouse (rm) IL-18 BPc and rmIL-18 BPd is observed.
Human
Videos

For Research Use Only