missing translation for 'onlineSavingsMsg'
Learn More

IL-16 Rat anti-Mouse, Clone: 228109, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1727

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438865

  • £376.32 / 500µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-16 Monoclonal antibody MAB1727 Specifically detects IL-16 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

IL-16
228109
Western Blot 1 ug/mL
O54824
FLJ16806, FLJ42735, FLJ44234, HsT19289, IL16, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCF, LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IL-16 in direct ELISAs and Western blots. Shows approximately 50% cross-reactivity with recombinant human (rh) IL-16 and no cross-reactivity with rmC-10, rhI-309, rhHCC-4, or rhHCC-1.
Mouse
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL16
Mouse myeloma cell line NS0-derived recombinant mouse IL-16 Ser1205-Ser1322 Accession # O54824
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3603
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only