Learn More
IL-15 R alpha Mouse anti-Human, Clone: 151303, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1471
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448665
  • £388.08 / 500µg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
IL-15 R alpha Monoclonal antibody MAB1471 Specifically detects IL-15 R alpha in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications
Specifications
IL-15 R alpha
151303
Purified
S. frugiperda insect ovarian cell linesf 21-derived recombinant human IL‑15 Rα
Protein A or G purified from hybridoma culture supernatant
RUO
Monoclonal
CyTOF
Unconjugated
Murine
IgG2b
500μg