missing translation for 'onlineSavingsMsg'
Learn More

IL-12 R beta 1 Mouse anti-Human, Clone: 69310, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB839

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13451916

  • £388.08 / 500µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-12 R beta 1 Monoclonal antibody MAB839 Specifically detects IL-12 R beta 1 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

IL-12 R beta 1
69310
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
P42701
CD212, CD212 antigen, IL-12 receptor beta component, IL-12 receptor subunit beta-1, IL12R, IL12R beta 1, IL-12R subunit beta-1, IL12RB, IL12RB1, IL-12RB1, IL-12R-beta-1, IL-12R-BETA1, interleukin 12 receptor, beta 1, interleukin-12 receptor beta-1 chain, interleukin-12 receptor subunit beta-1, MGC34454
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-12 R beta 1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-12 R beta 1 is observed.
Human
Flow Cytometry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL12RB1
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 1 Cys24-Glu540 Accession # P42701
Protein A or G purified from ascites
RUO
Primary
3594
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only