Learn More
IL-12/IL-23 p40 Mouse anti-Porcine, Clone: 116224, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB9121
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441986
  • £390.72 / 500µg
Estimated Shipment
10-09-2020
Description
Description
IL-12/IL-23 p40 Monoclonal antibody MAB9121 Specifically detects IL-12/IL-23 p40 in Porcine samples. It is validated for ELISA Capture
Specifications
Specifications
IL-12/IL-23 p40
116224
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Purified
Chinese hamster ovary cell line CHO-derived recombinant porcine IL-12 p70 heterodimer
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Detects the p40 subunit of porcine IL-12/IL-23 in ELISAs and Western blots. In ELISAs, this antibody shows 25-35% cross-reactivity with recombinant human IL-12 and IL-23 and no cross-reactivity with recombinant mouse IL-12 or IL-23.
Porcine
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Reconstitute at 0.5 mg/mL in sterile PBS.