missing translation for 'onlineSavingsMsg'
Learn More

IL-11 R alpha Mouse anti-Human, Clone: 473143, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1977

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13449075

  • £311.64 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-11R alpha Monoclonal antibody specifically detects IL-11R alpha in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-11R alpha
473143
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL11RA
Mouse myeloma cell line NS0-derived recombinant human IL-11 R alpha Cys26-Val363 Accession # Q14626
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3590
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q14626
AI314697, GP130, IL11R alpha, IL11RA, IL-11RA, Il11ra2, NR1
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-11 R alpha in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with rmIL-11 R alpha or recombinant human IL-6 R alpha is observed. In Western blots, 25% cross-reactivity with recombinant mouse (rm) IL-11 R alpha is observed.
Human
Videos
Documents

For Research Use Only