missing translation for 'onlineSavingsMsg'
Learn More

IL-10 R beta Mouse anti-Human, Clone: 90227, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB8741

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441946

  • £337.93 / 500µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IL-10 R beta Monoclonal antibody MAB8741 Specifically detects IL-10 R beta in Human samples. It is validated for Western Blotting
Specifications

Specifications

IL-10 R beta
90227
Western Blot 1 ug/mL
Q08334
CDw210b, CDw210b antigen, CRF2-4IL-10R subunit 2, CRFB4, CRFB4human transmembrane receptor protein; cytokine receptor10CDW210B, D21S58, D21S66, IBD25, IL-10 receptor subunit beta, IL10R beta, IL-10R2, IL-10R2IL-10R subunit beta, IL10RB, IL-10RB, interleukin 10 receptor, beta, Interleukin-10 receptor subunit 2, interleukin-10 receptor subunit beta, member 4
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human IL-10 R beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, ~10% cross-reactivity with recombinant human (rh) IL-10 R and rhIFN-alpha/beta R1 is observed and no cross-reactivity with rhIFN-gamma RI and rhIFN-gamma R2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL10RB
Mouse myeloma cell line NS0-derived recombinant human IL-10 R beta Met20-Ser220 Accession # Q08334
Protein A or G purified from ascites
RUO
Primary
3588
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos

For Research Use Only