Learn More
IL-10 Mouse anti-Canine, Clone: 138111, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB7351
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13441746
  • £364.33 / 500µg
Estimated Shipment
09-07-2019
Description
Description
IL-10 Monoclonal antibody MAB7351 Specifically detects IL-10 in Canine samples. It is validated for ELISA Capture
Specifications
Specifications
IL-10
138111
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
XP_855560
CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Delects canine IL-10 in ELISAs. In ELISAs, this antibody does not cross-react with recombinant feline IL-10, recombinant human (rh) IL-10, recombinant mouse (rm) IL-10, recombinant porcine IL-10, rhIL-10 sR, rmIL-10 sR, or recombinant rat IL-10.
Canine
ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IL10
E. coli-derived recombinant canine IL-10 Ser20-Ile179 Accession # XP_855560
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3586
Reconstitute at 0.5 mg/mL in sterile PBS.