Learn More
IL1R2 Monoclonal Antibody (34141), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA523820
Code : NEW
Additional Details : Weight : 0.00010kg
Description
When used in combination with the biotinylated human IL-1 RII affinity purified polyclonal detection antibody in sandwich ELISAs, no significant cross-reactivity or interference was observed with recombinant human (rh) IL-1ra, rhIL-1 RI, recombinant mouse IL-1ra, or recombinant rat IL-1ra. Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
IL1R2 Monoclonal antibody specifically detects IL1R2 in Human samples. It is validated for ELISA, Neutralization, Western Blot.Specifications
IL1R2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P27930 | |
Il1r2 | |
Sf 21-derived recombinant human IL-1 RII Phe14-Glu343 (Ser56Gly and Glu297Gly) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Neutralization, Western Blot | |
34141 | |
Unconjugated | |
Il1r2 | |
antigen CDw121b; CD121 antigen-like family member B; CD121b; CDw121b; IL-1 receptor beta chain; IL-1 type II receptor; Il1r2; Il-1r2; IL-1R-2; Il1r-2; IL1R2c; IL1RB; IL-1R-beta; IL-1RT2; IL-1RT-2; ILR2; interleukin 1 receptor type 2; interleukin 1 receptor type II variant 3; interleukin 1 receptor, type II; interleukin-1 receptor beta; Interleukin-1 receptor type 2; Interleukin-1 receptor type 2, membrane form; Interleukin-1 receptor type 2, soluble form; Interleukin-1 receptor type II; mIL-1R2; sIL-1R2; type II interleukin-1 receptor, beta | |
Mouse | |
Protein A/G | |
RUO | |
7850 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |