Learn More
IL-1 alpha/IL-1F1 Membrane Form Mouse anti-Human, Clone: 3405, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2001-100
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 15365467
  • £294.37 / 100µg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
IL-1 alpha/IL-1F1 Membrane Form Monoclonal antibody MAB2001100 Specifically detects IL-1 alpha/IL-1F1 Membrane Form in Human samples. It is validated for Flow Cytometry, Neutralization
Specifications
Specifications
IL-1 α/IL-1F1 Membrane Form
3405
Lyophilized from 0.2μm filtered solution in PBS with Trehalose.
Ascites
E. coli-derived recombinant human IL‑1α/IL‑1F1, Ser113-Ala271
Protein A or G purified from ascites
RUO
Monoclonal
Detects human IL‑1α/IL‑1F1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse IL-1α, recombinant rat IL-1α, recombinant cotton rat IL-1α, recombinant human (rh) IL-1β or rhIL-18 is observed.
CyTOF, Flow Cytometry, Neutralization
Unconjugated
P01583
Murine
IgG1
100μg
Primary
3552