missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-1 alpha/IL-1F1 Membrane Form Mouse anti-Human, Clone: 3405, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | IL-1 alpha/IL-1F1 |
---|---|
Clone | 3405 |
Applications | Flow Cytometry, Neutralization, CyTOF |
Conjugate | Unconjugated |
Form | Ascites |
Description
IL-1 alpha/IL-1F1 Monoclonal specifically detects IL-1 alpha/IL-1F1 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOF-ready.Specifications
IL-1 alpha/IL-1F1 | |
Flow Cytometry, Neutralization, CyTOF | |
Ascites | |
RUO | |
3552 | |
Monoclonal | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
3405 | |
Unconjugated | |
Mouse | |
Human | |
E. coli-derived recombinant human IL-1 alpha/IL-1F1 Ser113-Ala271 Accession # P01583 | |
Primary |
For Research Use Only