Learn More
IGSF2/CD101 Rat anti-Mouse, Clone: 307707, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3368
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13450036
  • £302.82 / 100µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
IGSF2/CD101 Monoclonal antibody MAB3368 Specifically detects IGSF2/CD101 in Mouse samples. It is validated for Flow Cytometry
Specifications
Specifications
IGSF2/CD101
307707
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD101
Chinese hamster ovary cell line CHO-derived recombinant mouse IGSF2/CD101 Gln21-Tyr1033 Accession # A8E0Y8
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
9398
Reconstitute at 0.5 mg/mL in sterile PBS.
Flow Cytometry, CyTOF
LYOPH
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
A8E0Y8
CD101, CD101 antigen, CD101 molecule, Cell surface glycoprotein V7, EWI101, EWI-101, Glu-Trp-Ile EWI motif-containing protein 101, IgSF2, Leukocyte Surface Protein, member 2, V7
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IGSF2/CD101.