missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgM κ, Rat, FITC, Clone: R4-22, Isotype Control, BD
Brand: BD Biosciences 553942
Additional Details : Weight : 0.01000kg
Description
The R4-22 antibody has unknown specificity.
Host Species: Rat
Clone: R4-22
Isotype: IgM, κ
Immunogen: Pooled Mouse Ig
Cytotoxicity Assay, Isotype Control
Specifications
Cytotoxicity Assay | |
0.5mg/mL | |
Rat | |
Affinity Purified | |
Undiluted at 4°C. Protect from exposure to light. Do not freeze. |
R4-22 | |
FITC | |
Pooled Mouse Ig | |
0.25 mg | |
IgM κ |