missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Fc Secondary Antibody, PE, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MA110381
Code : LD
Additional Details : Weight : 0.01000kg
Description
Human IgM Fc Monoclonal antibody specifically detects Human IgM Fc in Human samples. It is validated for Flow Cytometry.Specifications
Human IgM Fc | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Purified human IgM. | |
100 μg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
Flow Cytometry | |
CH2 | |
PE | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Liquid |