Learn More
IgM Mouse anti-Human, APC, Clone: G20-127, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561010
Additional Details : Weight : 0.01000kg
Description
The G20-127 monoclonal antibody specifically binds to the heavy chain of human IgM. The majority of mature B lymphocytes express IgM. The G20-127 antibody does not react with other immunoglobulin heavy chain isotypes.
Specifications
IgM | |
Monoclonal | |
APC | |
Mouse | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
G20-127 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |