Learn More
anti-IgG2a κ, Rat, R-PE, Clone: R35-95, Isotype Control, BD
Brand: BD Biosciences 551799
Additional Details : Weight : 0.01000kg
Description
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin
Intracellular Flow Cytometry
Specifications
Flow Cytometry | |
R-PE | |
Mouse Pooled Immunoglobulin | |
50 Tests | |
IgG2a κ |
R35-95 | |
Rat | |
Affinity Purified | |
Undiluted at 4°C |