Learn More
anti-IgG2a κ, Mouse, PE-Cy™7, Clone: G155-178, BD
Brand: BD Biosciences 557907
Additional Details : Weight : 0.01000kg
Description
The G155-178 clone has an unknown specificity. Trinitrophenal (TNP), the immunogen, is a hapten not expressed on human, mouse, rat or non-human primate cells. In the absence of specific binding, this antibody may bind non-specifically to Fc receptors. The immunoglobulin from clone G155-178 was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This immunoglobulin is specific for trinitrophenol (TNP), a hapten not expressed on human cells or human cell lines. In the absence of specific binding, this immunoglobulin may bind non-specifically to Fc receptors. This antibody is routinely tested on peripheral blood by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: G155-178
Isotype: IgG2a, κImmunogen: TNP-keyhole limpet hemocyanin
Flow Cytometry, Isotype Control
Specifications
Flow Cytometry | |
PE-Cyanine7 | |
TNP-keyhole limpet hemocyanin | |
100 Tests | |
IgG2a |
G155-178 | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |