Learn More
Mouse IgG1 κ, V450, Clone: MOPC-21, Isotype Control, BD
Brand: BD Biosciences 560373
262.71 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is
available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1, κ
Species Reactivity: Human
Flow Cytometry

Specifications
Flow Cytometry | |
0.2mg/mL | |
Mouse | |
0.1 mg | |
Undiluted at 4°C |
MOPC-21 | |
Violet Fluor 450 (V450) | |
Affinity Purified | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.