missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IgG1 κ, Mouse, FITC, Clone: MOPC-21, Isotype Control, BD
£289.00 - £313.00
Specifications
Clone | MOPC-21 |
---|---|
Applications | Flow Cytometry, Western Blot |
Conjugate | FITC |
Host Species | Mouse |
Isotype | IgG1 κ |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10014743
|
BD Biosciences
554679 |
0.1 mg |
£313.00
Each |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
15856738
|
BD Biosciences
551954 |
50 Tests |
£289.00
Each |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
MOPC-21 | |
FITC | |
IgG1 κ | |
Undiluted at 4°C |
Flow Cytometry, Western Blot | |
Mouse | |
Affinity Purified |