Learn More
anti-IgG1 κ, Mouse, FITC, Clone: MOPC-21, Isotype Control, BD
Brand: BD Biosciences 551954
Additional Details : Weight : 0.01000kg
Description
The MOPC-21 immunoglobulin is a mouse myeloma protein. The MOPC-21 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: MOPC-21
Isotype: IgG1 κ
Species Reactivity: Non-human primate
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
Flow Cytometry, Western Blot | |
FITC | |
Affinity Purified | |
Undiluted at 4°C |
MOPC-21 | |
Mouse | |
50 Tests | |
IgG1 κ |