Learn More
anti-IgG1 λ1, Hamster, Clone: G235-2356, Isotype Control, BD
Brand: BD Biosciences 554709
Additional Details : Weight : 0.01000kg
Description
The immunogen used to produce the G235-2356 hybridoma was the hapten trinitrophenol conjugated to a protein carrier. The G235-2356 antibody was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by functional assay. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Hamster
Clone: G235-2356
Isotype: IgG1, λ1
Immunogen: Trinitrophenol-KLH
Functional Assay, Isotype Control
Specifications
Functional Assay | |
1mg/mL | |
Hamster | |
0.5 mg | |
IgG1 λ1 |
G235-2356 | |
Unconjugated | |
Affinity Purified | |
Undiluted at 4°C |