missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545905
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AVVPLARTQT CGNCTFRVCF EQCCPWTFMV KLINQNCDSA RTSDDRLCRS Sequence homology: Human: 100%
IGFL1 Polyclonal antibody specifically detects IGFL1 in Human samples. It is validated for Western Blot.Specifications
IGFL1 | |
Polyclonal | |
Unconjugated | |
IGFL1 | |
APRG644; IGF; IGF like family member 1; IGFL1; IGF-like family member 1; Insulin growth factor-like family member 1; Insulin like growth factor; UNQ644; UNQ644/PRO1274 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
374918 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6UW32 | |
IGFL1 | |
Synthetic peptide directed towards the C-terminal of human IGFL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |