missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGFBP3 Monoclonal Antibody (84728.111), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120185
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-20185 detects a band of ∽41 kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
IGFBP3 Monoclonal antibody specifically detects IGFBP3 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot.Specifications
IGFBP3 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative | |
P17936 | |
IGFBP3 | |
Recombinant, full length, human IGFBP3 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
84728.111 | |
Unconjugated | |
IGFBP3 | |
acid stable subunit of the 140 K IGF complex; AI649005; binding protein 29; binding protein 53; BP53; BP-53; Growth hormone dependent binding protein; growth hormone-dependent binding protein; I79_011450; IBP3; IBP-3; IGF binding protein 3; IGF-binding protein 3; IGFBP; Igfbp3; IGF-BP3; IGFBP-3; IGgfbp3; Insulin like growth factor binding protein; insulin like growth factor binding protein 3; insulin-like growth factor binding protein 3; Insulin-like growth factor binding protein 3 precursor; insulin-like growth factor-binding protein (IGF-BP3); insulin-like growth factor-binding protein 3; tcag7.703 | |
Mouse | |
purified | |
RUO | |
3486 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |