missing translation for 'onlineSavingsMsg'
Learn More

IGFBP-6 Rat anti-Mouse, Clone: 122312, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB7761

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441856

  • £376.32 / 500µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

IGFBP-6 Monoclonal antibody MAB7761 Specifically detects IGFBP-6 in Mouse samples. It is validated for ELISA Capture, Western Blotting
Specifications

Specifications

IGFBP-6
122312
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P47880
IBP6, IBP-6, IGF binding protein 6, IGF-binding protein 6, IGFBP6, IGFBP-6, insulin-like growth factor binding protein 6, insulin-like growth factor-binding protein 6
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IGFBP-6 in ELISAs and Western blots. In Western blots, 10% cross-reactivity with recombinant human (rh) IGFBP-3 and rhIGFBP-5 is observed and no cross-reactivity with rhIGFBP-1, recombinant mouse (rm) IGFBP-1, rhIGFBP-2, rmIGFBP-2, rmIGFBP-3, rhIGFBP-4, rmIGFBP-5, or rhIGFBP-6 is observed.
Mouse
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IGFBP6
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-6 Gly24-Gly238 Accession # P47880
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3489
Reconstitute at 0.5 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only