Learn More
IGFBP-1 Rat anti-Mouse, Clone: 176110, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1240
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13438485
  • £405.72 / 500µg
Estimated Shipment date
from Supplier 16-12-2019
Description
Description
IGFBP-1 Monoclonal antibody MAB1240 Specifically detects IGFBP-1 in Mouse samples. It is validated for Western Blotting
Specifications
Specifications
IGFBP-1
176110
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
IGFBP1
Mouse myeloma cell line NS0-derived recombinant mouse IGFBP-1 Ala26-Asn272 Accession # P47876
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
3484
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P47876
alpha-pregnancy-associated endometrial globulin, amniotic fluid binding protein, binding protein-25, binding protein-26, binding protein-28, growth hormone independent-binding protein, hIGFBP-1, IBP1, IBP-1, IGF-binding protein 1, IGFBP1, IGFBP-1, IGF-BP25, insulin-like growth factor binding protein 1, insulin-like growth factor-binding protein 1, Placental protein 12, PP12AFBP
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse IGFBP-1 in direct ELISAs and Western blots. In Western blots, weak cross-reactivity with recombinant mouse (rm) IGFBP-5 is observed and no cross-reactivity with recombinant human (rh) IGFBP-1, rmIGFBP-2, rmIGFBP-3, rhIGFBP-4, rmIGFBP-6, or rhIGFBP-7 is observed.